Ethyl-4-(4-chlorophenyl)-4-oxobutyrate

Lieferant: Apollo Scientific

Synonyme: 4-(4-Chloro-phenyl)-4-oxo-butyric acid ethyl ester

OR307555-1G OR307555-5G
APOSOR307555-1GEA 343 EUR
APOSOR307555-1G APOSOR307555-5G
Ethyl-4-(4-chlorophenyl)-4-oxobutyrate
Ethyl-4-(4-chlorophenyl)-4-oxobutyrate
Formel: C₁₂H₁₃ClO₃
Molecular Weight: 240.69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00032767
CAS-Nummer: 53503-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR