6-(Cyclohexyloxy)pyridine-2-carboxylic acid

Lieferant: Apollo Scientific

OR30771-1G OR30771-50MG OR30771-250MG OR30771-500MG OR30771-2.5G
APOSOR30771-1GEA 335 EUR
APOSOR30771-1G APOSOR30771-50MG APOSOR30771-250MG APOSOR30771-500MG APOSOR30771-2.5G
6-(Cyclohexyloxy)pyridine-2-carboxylic acid
6-(Cyclohexyloxy)pyridine-2-carboxylic acid
Formel: C₁₂H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11100044
CAS-Nummer: 1215727-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR