6,7-Dihydro-2-(2,4,6-trimethylphenyl)-5H-pyrrolo[2,1-c]-1,2,4-triazolium perchlorate

Lieferant: Apollo Scientific

Synonyme: 6,7-Dihydro-2-mesityl-5H-pyrrolo[2,1-c]-1,2,4-triazolium Perchlorate

OR307710-1G OR307710-250MG
APOSOR307710-1GEA 723 EUR
APOSOR307710-1G APOSOR307710-250MG
6,7-Dihydro-2-(2,4,6-trimethylphenyl)-5H-pyrrolo[2,1-c]-1,2,4-triazolium perchlorate
6,7-Dihydro-2-(2,4,6-trimethylphenyl)-5H-pyrrolo[2,1-c]-1,2,4-triazolium perchlorate
Formel: C₁₄H₁₈ClN₃O₄
Molecular Weight: 327,77 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1334529-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR