7-Chlor-2,4-dimethylchinolin
Lieferant: Apollo Scientific
OR307822-1G
OR307822-5G
APOSOR307822-1GEA
293
EUR
APOSOR307822-1G
APOSOR307822-5G
7-Chlor-2,4-dimethylchinolin
7-Chloro-2,4-dimethylquinoline
Formel:
C₁₁H₁₀ClN Lagertemperatur: Raumtemperatur |
CAS-Nummer:
88499-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...