6-(4-Methylpiperazino)-3-pyridinamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-(4-methylpiperazin-1-yl)pyridine, 6-(4-Methylpiperazin-1-yl)pyridin-3-amine

OR30784-250MG
APOSOR30784-250MGEA 191 EUR
APOSOR30784-250MG
6-(4-Methylpiperazino)-3-pyridinamine
6-(4-Methylpiperazino)-3-pyridinamine
Formel: C₁₀H₁₆N₄
Molecular Weight: 192,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425719
CAS-Nummer: 55403-35-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR