4-Amino-2,6-dimethylchinolin
Lieferant: Apollo Scientific
OR307858-1G
OR307858-5G
APOSOR307858-1GEA
317
EUR
APOSOR307858-1G
APOSOR307858-5G
4-Amino-2,6-dimethylchinolin
4-Amino-2,6-dimethylquinoline
Formel:
C₁₁H₁₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01870544 CAS-Nummer: 342618-57-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...