4-Amino-6,8-dichlor-2-methylchinolin
Lieferant: Apollo Scientific
OR307860-1G
OR307860-5G
APOSOR307860-1GEA
317
EUR
APOSOR307860-1G
APOSOR307860-5G
4-Amino-6,8-dichlor-2-methylchinolin
4-Amino-6,8-dichloro-2-methylquinoline
Formel:
C₁₀H₈Cl₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669721 CAS-Nummer: 917562-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...