4-Amino-7,8-dichlor-2-methylchinolin

Lieferant: Apollo Scientific

OR307861-1G OR307861-5G
APOSOR307861-1GEA 277 EUR
APOSOR307861-1G APOSOR307861-5G
4-Amino-7,8-dichlor-2-methylchinolin
4-Amino-7,8-dichloro-2-methylquinoline
Formel: C₁₀H₈Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08669722
CAS-Nummer: 917562-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR