2,6-Dibromopyridin-4-amine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-2,6-dibromopyridine
OR30787-1G
OR30787-5G
OR30787-25G
APOSOR30787-1GEA
21.9
EUR
APOSOR30787-1G
APOSOR30787-5G
APOSOR30787-25G
2,6-Dibromopyridin-4-amine 95+%
2,6-Dibromopyridin-4-amine
Formel:
C₅H₄Br₂N₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD00234047 CAS-Nummer: 39771-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...