2,5,8-Trimethyl-3-chinolincarbonsäureethylester
Lieferant: Apollo Scientific
OR307879-1G
OR307879-5G
APOSOR307879-1GEA
293
EUR
APOSOR307879-1G
APOSOR307879-5G
2,5,8-Trimethyl-3-chinolincarbonsäureethylester
2,5,8-Trimethylquinoline-3-carboxylic acid ethyl ester
Formel:
C₁₅H₁₇NO₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
110139-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...