2,6,8-Trimethyl-3-chinolincarbonsäureethylester

Lieferant: Apollo Scientific

OR307883-1G OR307883-5G
APOSOR307883-1GEA 293 EUR
APOSOR307883-1G APOSOR307883-5G
2,6,8-Trimethyl-3-chinolincarbonsäureethylester
2,6,8-Trimethylquinoline-3-carboxylic acid ethyl ester
Formel: C₁₅H₁₇NO₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 948291-48-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR