2-Chlor-6-ethoxy-3-methylchinolin
Lieferant: Apollo Scientific
OR307918-1G
OR307918-5G
APOSOR307918-1GEA
293
EUR
APOSOR307918-1G
APOSOR307918-5G
2-Chlor-6-ethoxy-3-methylchinolin
2-Chloro-6-ethoxy-3-methylquinoline
Formel:
C₁₂H₁₂ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787670 CAS-Nummer: 948291-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...