2-Chloro-7,8-dimethylquinoline-3-carbonitrile

Lieferant: Apollo Scientific

OR307924-1G OR307924-5G
APOSOR307924-1GEA 301 EUR
APOSOR307924-1G APOSOR307924-5G
2-Chloro-7,8-dimethylquinoline-3-carbonitrile
2-Chloro-7,8-dimethylquinoline-3-carbonitrile
Formel: C₁₂H₉ClN₂
Molecular Weight: 216.67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09035783
CAS-Nummer: 917746-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR