4,6-Dichlor-8-methylchinolin

Lieferant: Apollo Scientific

OR307931-1G OR307931-5G
APOSOR307931-1GEA 169 EUR
APOSOR307931-1G APOSOR307931-5G
4,6-Dichlor-8-methylchinolin
4,6-Dichloro-8-methylquinoline
Formel: C₁₀H₇Cl₂N
Lagertemperatur: Raumtemperatur
CAS-Nummer: 948292-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR