4-Amino-7-chlorchinolin 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
7-Chloroquinolin-4-amine
OR307952-1G
OR307952-5G
APOSOR307952-1GEA
72.5
EUR
APOSOR307952-1G
APOSOR307952-5G
4-Amino-7-chlorchinolin 95%
4-Amino-7-chloroquinoline
Formel:
C₉H₇ClN₂ Molecular Weight: 178,6183 g/mol Schmelzpunkt: 143…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00828822 CAS-Nummer: 1198-40-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...