4-Amino-8-chlorchinolin
Lieferant: Apollo Scientific
OR307957-1G
OR307957-5G
APOSOR307957-1GEA
368
EUR
APOSOR307957-1G
APOSOR307957-5G
4-Amino-8-chlorchinolin
4-Amino-8-chloroquinoline
Formel:
C₉H₇ClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037192 CAS-Nummer: 65340-72-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...