4-Amino-8-methoxy-2-methylchinolin

Lieferant: Apollo Scientific

OR307958-1G OR307958-250MG
APOSOR307958-1GEA 348 EUR
APOSOR307958-1G APOSOR307958-250MG
4-Amino-8-methoxy-2-methylchinolin
4-Amino-8-methoxy-2-methylquinoline
Formel: C₁₁H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08754055
CAS-Nummer: 657391-86-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR