4-Chlor-2,5,7-trimethylchinolin

Lieferant: Apollo Scientific

OR307960-1G OR307960-5G
APOSOR307960-1GEA 301 EUR
APOSOR307960-1G APOSOR307960-5G
4-Chlor-2,5,7-trimethylchinolin
4-Chloro-2,5,7-trimethylquinoline
Formel: C₁₂H₁₂ClN
Lagertemperatur: Raumtemperatur
CAS-Nummer: 63136-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR