4-Chlor-2,6,8-trimethylchinolin
Lieferant: Apollo Scientific
OR307961-1G
OR307961-5G
APOSOR307961-1GEA
302
EUR
APOSOR307961-1G
APOSOR307961-5G
4-Chlor-2,6,8-trimethylchinolin
4-Chloro-2,6,8-trimethylquinoline
Formel:
C₁₂H₁₂ClN Lagertemperatur: Raumtemperatur |
CAS-Nummer:
87602-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...