4-Chlor-6,8-dimethylchinolin

Lieferant: Apollo Scientific

OR307965-1G
APOSOR307965-1GEA 95.7 EUR
APOSOR307965-1G
4-Chlor-6,8-dimethylchinolin
4-Chloro-6,8-dimethylquinoline
Formel: C₁₁H₁₀ClN
Molecular Weight: 191,6568 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00087699
CAS-Nummer: 196803-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR