4-Chlor-6-ethoxychinolin
Lieferant: Apollo Scientific
OR307967-1G
OR307967-5G
APOSOR307967-1GEA
325
EUR
APOSOR307967-1G
APOSOR307967-5G
4-Chlor-6-ethoxychinolin
4-Chloro-6-ethoxyquinoline
Formel:
C₁₁H₁₀ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787733 CAS-Nummer: 103862-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...