4-Chlor-8-methoxychinolin

Lieferant: Apollo Scientific

OR307972-5G
APOSOR307972-5GEA 139 EUR
APOSOR307972-5G
4-Chlor-8-methoxychinolin
4-Chloro-8-methoxyquinoline
Formel: C₁₀H₈ClNO
Molecular Weight: 193,6296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00033679
CAS-Nummer: 16778-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR