6,8-Dichlor-4-chinolinol

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxy-6,8-Dichloroquinoline

OR307976-1G OR307976-250MG
APOSOR307976-1GEA 261 EUR
APOSOR307976-1G APOSOR307976-250MG
6,8-Dichlor-4-chinolinol
6,8-Dichlor-4-chinolinol
Formel: C₉H₅Cl₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00272390
CAS-Nummer: 57935-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR