tert-Butyl-4-aminobutyl(methyl)carbamate

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-aminobutyl(methyl)carbamate, N-(4-Aminobutyl)-N-methyl carbamic acid tert-butyl ester

OR308002-1G
APOSOR308002-1GEA 144 EUR
APOSOR308002-1G
tert-Butyl-4-aminobutyl(methyl)carbamate
tert-Butyl-4-aminobutyl(methyl)carbamate
Formel: C₁₀H₂₂N₂O₂
Molecular Weight: 202,2900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001681
CAS-Nummer: 144222-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR