3-Azetidinone hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Azetidin-3-one hydrochloride

OR308018-25G
APOSOR308018-25GEA 75.4 EUR
APOSOR308018-25G
3-Azetidinone hydrochloride 95%
3-Azetidinone hydrochloride
Formel: C₃H₆ClNO
Molecular Weight: 107,54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01861742
CAS-Nummer: 17557-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR