3-Methylazetidin-3-ol hydrochloride
Lieferant: Apollo Scientific
OR308021-1G
OR308021-5G
APOSOR308021-1GEA
74.1
EUR
APOSOR308021-1G
APOSOR308021-5G
3-Methylazetidin-3-ol hydrochloride
3-Methylazetidin-3-ol hydrochloride
Formel:
C4H10ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100994 CAS-Nummer: 124668-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...