1-Methylazetidin-3-amine
Lieferant: Apollo Scientific
|
Gefahr
|
OR308028-1G
OR308028-100MG
OR308028-250MG
APOSOR308028-1GEA
591
EUR
APOSOR308028-1G
APOSOR308028-100MG
APOSOR308028-250MG
1-Methylazetidin-3-amine
1-Methylazetidin-3-amine
Formel:
C₄H₁₀N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11849029 CAS-Nummer: 959957-92-7 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...