3-(2-Aminoethyl)-1-boc-azetidine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate, 3-(1-Aminoéthyl)-1-azétidinecarboxylate de 2-méthyl-2-propanyle, 1-Boc-(3-Aminoethyl)azetidine, 1-Azetidinecarboxylic acid, 3-(1-aminoethyl)-, 1,1-dimethylethyl ester

OR308034-1G OR308034-250MG
APOSOR308034-1GEA 98.6 EUR
APOSOR308034-1G APOSOR308034-250MG
3-(2-Aminoethyl)-1-boc-azetidine 97%
3-(2-Aminoethyl)-1-boc-azetidine
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,282 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878606
CAS-Nummer: 898271-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR