5-Amino-7-azaindole

Lieferant: Apollo Scientific

Synonyme: 1H-Pyrrolo[2,3-b]pyridin-5-amine

OR308039-1G OR308039-5G
APOSOR308039-1GEA 88.6 EUR
APOSOR308039-1G APOSOR308039-5G
5-Amino-7-azaindole
5-Amino-7-azaindole
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659683
CAS-Nummer: 100960-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR