6-Amino-1,3-benzoxazol-2(3H)-one 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-2-hydroxybenzoxazole, 6-Amino-2-benzoxazolone, 6-Amino-2-benzoxazolinone

OR30804-5G
APOSOR30804-5GEA 236 EUR
APOSOR30804-5G
6-Amino-1,3-benzoxazol-2(3H)-one 96%
6-Amino-1,3-benzoxazol-2(3H)-one
Formel: C₇H₆N₂O₂
Molecular Weight: 150,14 g/mol
Schmelzpunkt: 199…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00463899
CAS-Nummer: 22876-17-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR