5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-7-azaindole-3-carboxylic acid
OR308043-1G
OR308043-250MG
OR308043-5G
APOSOR308043-1GEA
95.7
EUR
APOSOR308043-1G
APOSOR308043-250MG
APOSOR308043-5G
5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
Formel:
C₈H₅BrN₂O₂ Molecular Weight: 241,0400 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09965908 CAS-Nummer: 849068-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...