4-Chloro-2-iodo-7-azaindole

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-iodo-1H-pyrrolo[2,3-b]pyridine

OR308046-1G
APOSOR308046-1GEA 220 EUR
APOSOR308046-1G
4-Chloro-2-iodo-7-azaindole
4-Chloro-2-iodo-7-azaindole
Formel: C₇H₄ClIN₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 940948-29-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR