3-Bromo-4-chloro-7-azaindole

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine

OR308047-10G OR308047-5G OR308047-25G
APOSOR308047-10GEA 183 EUR
APOSOR308047-10G APOSOR308047-5G APOSOR308047-25G
3-Bromo-4-chloro-7-azaindole
3-Bromo-4-chloro-7-azaindole
Formel: C7H4BrClN2
Molecular Weight: 231.4800 g/mol
Schmelzpunkt: 214…215 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09880132
CAS-Nummer: 1000340-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR