1H-Indazol-4-boronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-1H-indazole, 1H-indazol-4-boronic acid pinacol ester

OR308052-5G
APOSOR308052-5GEA 122 EUR
APOSOR308052-5G
1H-Indazol-4-boronsäurepinakolester
1H-Indazol-4-boronsäurepinakolester
Formel: C₁₃H₁₇BN₂O₂
Molecular Weight: 244,10 g/mol
Schmelzpunkt: 139…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08059267
CAS-Nummer: 885618-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR