(R)-1-Boc-2-(Hydroxymethyl)piperazine

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-tert-Butyl 2-(hydroxymethyl)piperazine-1-carboxylate, tert-Butyl (2R)-2-(hydroxymethyl)piperazine-1-carboxylate

OR308074-5G
APOSOR308074-5GEA 192 EUR
APOSOR308074-5G
(R)-1-Boc-2-(Hydroxymethyl)piperazine
(R)-1-Boc-2-(Hydroxymethyl)piperazine
Formel: C₁₀H₂₀N₂O₃
Molecular Weight: 216,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772092
CAS-Nummer: 169448-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR