[1,2,4]Triazolo[4,3-a]pyridine-6-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR308081-1G OR308081-5G
APOSOR308081-1GEA 112 EUR
APOSOR308081-1G APOSOR308081-5G
[1,2,4]Triazolo[4,3-a]pyridine-6-carboxylic acid
[1,2,4]Triazolo[4,3-a]pyridine-6-carboxylic acid
Formel: C₇H₅N₃O₂
Molecular Weight: 163,1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07376943
CAS-Nummer: 933708-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR