3H-Imidazo[4,5-c]pyridine-4-carboxylic acid 97%

Lieferant: Apollo Scientific

OR308082-1G OR308082-500MG OR308082-5G
APOSOR308082-1GEA 725 EUR
APOSOR308082-1G APOSOR308082-500MG APOSOR308082-5G
3H-Imidazo[4,5-c]pyridine-4-carboxylic acid 97%
3H-Imidazo[4,5-c]pyridine-4-carboxylic acid
Formel: C₇H₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18803553
CAS-Nummer: 933728-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR