3-Aminocyclobutanone hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-aminocyclobutan-1-one hydrochloride

OR308095-10G OR308095-5G
APOSOR308095-10GEA 171 EUR
APOSOR308095-10G APOSOR308095-5G
3-Aminocyclobutanone hydrochloride 97%
3-Aminocyclobutanone hydrochloride
Formel: C₄H₈ClNO
Molecular Weight: 121,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12923220
CAS-Nummer: 1035374-20-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR