cis-tert-Butyl-3-hydroxycyclobutylcarbamate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl (cis-3-hydroxycyclobutyl)carbamate

OR308102-1G OR308102-5G OR308102-25G
APOSOR308102-1GEA 84.1 EUR
APOSOR308102-1G APOSOR308102-5G APOSOR308102-25G
cis-tert-Butyl-3-hydroxycyclobutylcarbamate 97%
cis-tert-Butyl-3-hydroxycyclobutylcarbamate
Formel: C₉H₁₇NO₃
Molecular Weight: 187,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038208
CAS-Nummer: 389890-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR