4-Phenyl-tetrahydro-pyran-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Phenyltetrahydro-2H-pyran-4-carboxylic acid

OR30811-1G OR30811-250MG
APOSOR30811-1GEA 116 EUR
APOSOR30811-1G APOSOR30811-250MG
4-Phenyl-tetrahydro-pyran-4-carboxylic acid
4-Phenyl-tetrahydro-pyran-4-carboxylic acid
Formel: C₁₂H₁₄O₃
Molecular Weight: 206,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085758
CAS-Nummer: 182491-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR