1-Boc-4-amino-4-methylpiperidine

Lieferant: Apollo Scientific

Synonyme: 1-Boc-4-Amino-4-methylpiperidine, tert-Butyl 4-amino-4-methylpiperidine-1-carboxylate

OR308122-1G OR308122-5G
APOSOR308122-1GEA 91.5 EUR
APOSOR308122-1G APOSOR308122-5G
1-Boc-4-amino-4-methylpiperidine
1-Boc-4-amino-4-methylpiperidine
Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214,3100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03265497
CAS-Nummer: 343788-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR