(S)-Morpholin-2-ylmethanol hydrochloride

Lieferant: Apollo Scientific

OR308169-5G
APOSOR308169-5GEA 77 EUR
APOSOR308169-5G
(S)-Morpholin-2-ylmethanol hydrochloride
(S)-Morpholin-2-ylmethanol hydrochloride
Formel: C₅H₁₂ClNO₂
Molecular Weight: 153,6100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13191722
CAS-Nummer: 1313584-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR