Boc-(4-carboxymethyl)piperazine

Lieferant: Apollo Scientific
Achtung

Synonyme: [1-(tert-Butoxycarbonyl)piperazin-4-yl]acetic acid, 2-(4-(tert-Butoxycarbonyl)piperazin-1-yl)acetic acid

OR30823-25G
APOSOR30823-25GEA 154 EUR
APOSOR30823-25G
Boc-(4-carboxymethyl)piperazine
Boc-(4-carboxymethyl)piperazine
Formel: C₁₁H₂₀N₂O₄
Molecular Weight: 244,29 g/mol
Schmelzpunkt: 186,5…189.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682402
CAS-Nummer: 156478-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR