Methyl 1H-pyrazole-4-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR30826-1G
OR30826-250MG
OR30826-5G
APOSOR30826-1GEA
27.7
EUR
APOSOR30826-1G
APOSOR30826-250MG
APOSOR30826-5G
Methyl 1H-pyrazole-4-carboxylate 97%
Methyl 1H-pyrazole-4-carboxylate
Formel:
C₅H₆N₂O₂ Molecular Weight: 126,113 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00297363 CAS-Nummer: 51105-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...