Methyl-6-chlor-5-nitronicotinat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 6-chloro-5-nitronicotinate
OR30827-1G
OR30827-25G
OR30827-5G
APOSOR30827-1GEA
29
EUR
APOSOR30827-1G
APOSOR30827-25G
APOSOR30827-5G
Methyl-6-chlor-5-nitronicotinat 97%
Methyl-6-chlor-5-nitronicotinat
Formel:
C₇H₅ClN₂O₄ Molecular Weight: 216,58 g/mol Schmelzpunkt: 76 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01928341 CAS-Nummer: 59237-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...