6-(Bromomethyl)-1H-indazole hydrobromide

Lieferant: Apollo Scientific
Achtung

OR30833-250MG OR30833-100MG
APOSOR30833-250MGEA 136 EUR
APOSOR30833-250MG APOSOR30833-100MG
6-(Bromomethyl)-1H-indazole hydrobromide
6-(Bromomethyl)-1H-indazole hydrobromide
Formel: C8H8Br2N2
Molecular Weight: 291,97 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD14586465
CAS-Nummer: 368426-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR