3-(3-Bromphenyl)propionsäure 97%
Lieferant: Apollo Scientific
OR3084-100G
APOSOR3084-100GEA
163
EUR
APOSOR3084-100G
3-(3-Bromphenyl)propionsäure 97%
3-(3-Bromphenyl)propionsäure
Formel:
C₉H₉BrO₂ Molecular Weight: 229,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310792 CAS-Nummer: 42287-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...