1-(1H-Indazol-5-yl)ethanone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Acetyl-1H-indazole
OR30885-1G
OR30885-500MG
APOSOR30885-1GEA
329
EUR
APOSOR30885-1G
APOSOR30885-500MG
1-(1H-Indazol-5-yl)ethanone
1-(1H-Indazol-5-yl)ethanone
Formel:
C₉H₈N₂O Molecular Weight: 160,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11869763 CAS-Nummer: 1001906-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...