3-Methoxy-3-methylazetidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR309010-1G
OR309010-5G
OR309010-25G
APOSOR309010-1GEA
89.9
EUR
APOSOR309010-1G
APOSOR309010-5G
APOSOR309010-25G
3-Methoxy-3-methylazetidine hydrochloride
3-Methoxy-3-methylazetidine hydrochloride
Formel:
C₅H₁₂ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100995 CAS-Nummer: 905843-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...