tert-Butyl-2-(aminomethyl)azetidine-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, tert-Butyl 2-(aminomethyl)azetidine-1-carboxylate

OR309032-1G OR309032-250MG
APOSOR309032-1GEA 505 EUR
APOSOR309032-1G APOSOR309032-250MG
tert-Butyl-2-(aminomethyl)azetidine-1-carboxylate
tert-Butyl-2-(aminomethyl)azetidine-1-carboxylate
Formel: C₉H₁₈N₂O₂
Molecular Weight: 186.26 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08544502
CAS-Nummer: 939760-37-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR